Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Morpholine 99+% ACS

Glass bottle for solids, 500 g
Catalog # ACRO415165000
Toxic
Corrosive
Flammable
Morpholine 99+% ACS
Catalog # ACRO415165000
CAS Number:  110-91-8
Morpholine 99+% ACS
Morpholine 99+% ACS

Glass bottle for solids, 500 g

CAS Number:  110-91-8
Price
53,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:12,06362
  • Boiling Pt:128.9 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:0.99 g/cm³
  • Flash Pt:32 °C
  • Storage Temperature:Ambient
  • EINECS:203-815-1
  • UN:2054
  • ADR:8,I

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4550 (20°C, 589 nm)
  • Boiling point:
    126.0 to 130.0°C
  • Color scale:
    ≤15 APHA
  • Titration with HCl:
    ≥99.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications