Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Bulb pipettes, BLAUBRAND®, two marks, class AS

Bulb pipettes, BLAUBRAND®, two marks, class AS

10 ml, 0,02 ± ml, 440 mm, Red
Catalogus # 612-2731
Leverancier:   Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Catalogus # 612-2731
Leverancier:  Brand
Bulb pipettes, BLAUBRAND®, two marks, class AS
Bulb pipettes, BLAUBRAND®, two marks, class AS

10 ml, 0,02 ± ml, 440 mm, Red

Price
11,10 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including one batch certificate

DIN EN ISO 648

Certifications: DIN EN ISO 648; DE-M marking.

Specificaties
  • Colour code:
    Red
  • Capacity:
    10 ml
  • Length:
    440 mm
  • Seq:
    06
  • Tolerance:
    0,02 ± ml
Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.