Borane trimethylamine complex 97%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Borane trimethylamine complex 97%
Catalog # ACRO402741000
Supplier: Thermo Fisher Scientific
CAS Number: 75-22-9
Jump to:
Product Details & Documents
- CAS Number:75-22-9
- Formula:C₃H₁₂BN
- MDL Number:MFCD00012420
- Molecular Weight:72.95 g/mol
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
- EINECS:200-850-4
- UN:1325
- ADR:4.1,II
Documents
ACRO40274
SDS
Specifications
Specification Test Results
- Appearance:White Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:92 to 96 °C
- Titration with Na2S2O3:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



