Laboratory trays
290×225×50 mm, 260×190×50 mm
Catalogus # 216-1362
Leverancier: BOCHEM
Ga naar:
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
- Rounded corners for fast, easy cleaning
- Polished electrolytically, anti-magnetic
- Good chemical resistance, excellent thermal resistance
Multipurpose trays for a wide range of uses. Ideal as developing trays.
Specificaties
- W×D×H ext.:290×225×50 mm
- Seq:03
- W×D×H int.:260×190×50 mm
Aanbevelingen