Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sodium taurocholate hydrate 98%

Sodium taurocholate hydrate 98%

Glass bottle, 5 g
Catalogus # ACRO230370050
Irriterend
Leverancier:   Thermo Fisher Scientific
Sodium taurocholate hydrate 98%
Catalogus # ACRO230370050
CAS-nummer:  345909-26-4
Sodium taurocholate hydrate 98%
Sodium taurocholate hydrate 98%

Glass bottle, 5 g

CAS-nummer:  345909-26-4
Price
180,00 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
Synoniemen:  Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate
  • CAS-nummer:345909-26-4
  • Formule:C₂₆H₄₆NNaO₈S
  • MDL-nummer:MFCD00150819
  • Molecular Weight:555.70 g/mol
  • Smelten Pt:230 °C
  • Temperatuur bij opslag:Ambient

Documents

Specificaties

Specificatie testresultaten

  • Appearance:
    White to cream Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤7 %
  • TLC:
    ≥97.5 %
  • Residue after ignition:
    11.0 % to 14.0 %
  • Specific optical rotation:
    +20.0° to +25.8° (20°C, 589 nm) (c=3, water)
  • Heavy metals (as Pb):
    ≤20 ppm

Specificaties

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specificaties

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.