D-(-)-Glutamic acid 99+%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
D-(-)-Glutamic acid 99+%
Catalog # ACRO227891000
Supplier: Thermo Fisher Scientific
CAS Number: 6893-26-1
Jump to:
Product Details & Documents
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:200…202 °C
- Density:1,533 g/cm³ (20 °C)
- EINECS:230-000-8
Documents
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≤1 % L-content
- Specific optical rotation:-30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with NaOH:99.0 to 101.0 %
- Other amino acids:≤0.5 % (TLC)
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



