D-(-)-Glutamic acid 99+%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
D-(-)-Glutamic acid 99+%
Catalog # ACRO227890250
Supplier: Thermo Fisher Scientific
CAS Number: 6893-26-1
Jump to:
Product Details & Documents
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:200…202 °C
- Density:1,533 g/cm³ (20 °C)
- EINECS:230-000-8
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≤1 % L-content
- Specific optical rotation:-30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with NaOH:99.0 to 101.0 %
- Other amino acids:≤0.5 % (TLC)
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



