Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

D-(-)-Glutamic acid 99+%

Glass bottle for solids, 5 g
Catalog # ACRO227890050
D-(-)-Glutamic acid 99+%
Catalog # ACRO227890050
CAS Number:  6893-26-1
D-(-)-Glutamic acid 99+%
D-(-)-Glutamic acid 99+%

Glass bottle for solids, 5 g

CAS Number:  6893-26-1
Price
48,40 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:200…202 °C
  • Density:1,533 g/cm³ (20 °C)
  • EINECS:230-000-8
Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≤1 % L-content
  • Specific optical rotation:
    -30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
  • Titration with NaOH:
    99.0 to 101.0 %
  • Other amino acids:
    ≤0.5 % (TLC)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications