CD10 (FR4D11) mouse monoclonal IgG1 200 µg/ml raised against Raji cells 1 * 0,5 mL
SANTSC-7632
CD10 (FR4D11) mouse monoclonal IgG1 200 µg/ml raised against Raji cells 1 * 0,5 mL
SANTSC-7632
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK