IFN-alpha/betaRalpha (H-11) mouse monoclonal IgG1 200 µg/ml raised against amino acids 458-557 mapping at the C-terminus (complet e intracellular domain) of the IFN-&alpha /&beta R alpha chain precursor of human origin 1 * 1 mL
Catalog # SANTSC-7391
Supplier: Santa Cruz Biotechnology
Frequently Bought Together