Order Entry
Netherlands
Orders LinkContactUsLinkComponent
pH Buffer solutions in bottles

pH Buffer solutions in bottles

pH Buffer solution, 4,01, ±0,01, Colourless, 25 °C, 500 mL
Catalog # HANNHI7004L
Supplier:   Hanna
pH Buffer solutions in bottles
Catalog # HANNHI7004L
Supplier:  Hanna
CAS Number:  
pH Buffer solutions in bottles

pH Buffer solution, 4,01, ±0,01, Colourless, 25 °C, 500 mL

CAS Number:  
Price
33,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Plastic bottle with a tamper-proof seal.

  • Color coded label

Traceable to NIST standard reference materials.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • Description:
    pH Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.