Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Forceps, simple type
Forceps, simple type
Straight, sharp, serrated, 105 mm
Catalog # HAMMHSC800-10
Supplier:   HAMMACHER KARL
Forceps, simple type
Catalog # HAMMHSC800-10
Supplier:  HAMMACHER KARL
Forceps, simple type
Forceps, simple type
Straight, sharp, serrated, 105 mm
Price
22,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Sharp forceps made from stainless steel.
Specifications
  • Type:
    Straight, sharp, serrated
  • Length:
    105 mm
  • Seq:
    01