Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Reversed phase bulk packing material

Reversed phase bulk packing material

PRP-1 bulk resin, 5 µm, 100 Å, 1 g
Catalog # 554-1328
Supplier:   HAMILTON BONADUZ
Reversed phase bulk packing material
Catalog # 554-1328
Supplier:  HAMILTON BONADUZ
CAS Number:  
Reversed phase bulk packing material
Reversed phase bulk packing material

PRP-1 bulk resin, 5 µm, 100 Å, 1 g

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

Polymer bulk packing material for reversed phase applications.

Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.

Documents

Specifications

Specifications

  • Particle Size:
    5 µm
  • Pk:
    1 g
  • Pore size:
    100 Å
  • Description:
    PRP-1 bulk resin

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.