BUFFER SOLUTION PH 4.01 25C 1 * 500 mL
Catalog # REGEEP401
Supplier: REAGECON
Frequently Bought Together
Accessories for pH/°C meter, handheld
Supplier: Testo
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonium sulfate EMPROVE EXPERT ChP,NF,ACS 1 * 1 kg
Supplier: MERCK PRODUCTION CHEMICALS
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Reference standard kits for use with MD 100 and MD 200 photometers
Supplier: Lovibond
CONDUCTIVITY STANDARDS 5µS/CM 1 * 250 mL
Supplier: Mettler - Toledo
Disposable cuvettes
Supplier: Brand
Tubing valves, LaboPlast®
Supplier: BURKLE
Redox buffer solutions
Supplier: Mettler - Toledo
BUFFER SOLUTION PH 6.87 25C 1 * 500 mL
Supplier: REAGECON