Enniatin Complex
Bottle
Catalog # APOSBIE9701-100MG
New Product
Supplier: Apollo Scientific
Enniatin Complex
Catalog # APOSBIE9701-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 11113-62-5
Jump to:
Product Details & Documents
- CAS Number:11113-62-5
- MDL Number:MFCD00214209
- Melting Pt:120…128 °C
- Melting Pt:120-128 °C
- Storage Temperature:Freezer
- UN:3462
- ADR:6.1,III
Specifications
Specifications
- Pack type:Bottle



