Enniatin Complex
Bottle
Catalog # APOSBIE9701-50MG
New Product
Supplier: Apollo Scientific
Enniatin Complex
Catalog # APOSBIE9701-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 11113-62-5
Jump to:
Product Details & Documents
- CAS Number:11113-62-5
- MDL Number:MFCD00214209
- Melting Pt:120…128 °C
- Melting Pt:120-128 °C
- Storage Temperature:Freezer
- UN:3462
- ADR:6.1,III
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



