Order Entry
Netherlands
Orders LinkContactUsLinkComponent
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

Bottle
Catalog # APOSOR919237-25G
New Product
Supplier:   Apollo Scientific
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
Catalog # APOSOR919237-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  66185-74-8
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%

Bottle

CAS Number:  66185-74-8
Price
916,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:66185-74-8
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD10566907
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications