Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle
Catalog # APOSOR937522-100G
New Product
Supplier:   Apollo Scientific
Methyl trans-4-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR937522-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  6125-57-1
Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle

CAS Number:  6125-57-1
Price
482,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate
  • CAS Number:6125-57-1
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD20441963
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.