Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle
Catalog # APOSOR937522-25G
New Product
Supplier:   Apollo Scientific
Methyl trans-4-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR937522-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  6125-57-1
Methyl trans-4-hydroxycyclohexanecarboxylate 95%

Bottle

CAS Number:  6125-57-1
Price
122,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate
  • CAS Number:6125-57-1
  • Formula:C₈H₁₄O₃
  • MDL Number:MFCD20441963
  • Molecular Weight:158.20 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications