Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

[3-(Methylamino)phenyl]methanol

Bottle
Catalog # APOSOR310887-1G
New Product
Supplier:   Apollo Scientific
[3-(Methylamino)phenyl]methanol
Catalog # APOSOR310887-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  121562-78-5
[3-(Methylamino)phenyl]methanol

Bottle

CAS Number:  121562-78-5
Price
1 230,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:121562-78-5
  • Formula:C₈H₁₁NO
  • MDL Number:MFCD16658585
  • Molecular Weight:137.179 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications