Morpholine 99%
Bottle
Catalog # APOSOR13977-2.5KG
New Product
Supplier: Apollo Scientific
Morpholine 99%
Catalog # APOSOR13977-2.5KG
Supplier: Apollo Scientific
New Product
CAS Number: 110-91-8
Jump to:
Product Details & Documents
- CAS Number:110-91-8
- Formula:C₄H₉NO
- MDL Number:MFCD00005972
- Molecular Weight:87.12 g/mol
- Boiling Pt:129 °C
- Melting Pt:–7…–5 °C
- Melting Pt:–7 to –5 °C
- Density:0.996 g/cm³ (25 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-815-1
- UN:2054
- ADR:8+3,I
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



