Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Nipecotamide

Bottle
Catalog # APOSOR13974-500G
Irritant
New Product
Supplier:   Apollo Scientific
Nipecotamide
Catalog # APOSOR13974-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  4138-26-5
Nipecotamide

Bottle

CAS Number:  4138-26-5
Price
372,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Piperidine-3-carboxamide
  • CAS Number:4138-26-5
  • Formula:C₆H₁₂N₂O
  • MDL Number:MFCD00005993
  • Molecular Weight:128.1700 g/mol
  • Melting Pt:103…106 °C
  • Storage Temperature:Ambient
  • EINECS:223-962-5
Specifications

Specifications

  • Pack type:
    Bottle

Specifications