Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Nipecotamide

Nipecotamide

Bottle
Catalog # APOSOR13974-100G
Irritant
New Product
Supplier:   Apollo Scientific
Nipecotamide
Catalog # APOSOR13974-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  4138-26-5
Nipecotamide

Bottle

CAS Number:  4138-26-5
Price
88,30 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Piperidine-3-carboxamide
  • CAS Number:4138-26-5
  • Formula:C₆H₁₂N₂O
  • MDL Number:MFCD00005993
  • Molecular Weight:128.1700 g/mol
  • Melting Pt:103…106 °C
  • Storage Temperature:Ambient
  • EINECS:223-962-5
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.