Order Entry
Netherlands
Orders LinkContactUsLinkComponent
AR42 ≥98%

AR42 ≥98%

New Product
Supplier:   Apollo Scientific
AR42 ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  HDAC-42, (S)-N-Hydroxy-4-(3-methyl-2-phenylbutanamido)benzamide
  • CAS Number:935881-37-1
  • Formula:C₁₈H₂₀N₂O₃
  • Molecular Weight:312,36 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 335,00 €
    335,00 €
    1 * 25 mg
       335,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 545,00 €
    545,00 €
    1 * 50 mg
       545,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 831,00 €
    831,00 €
    1 * 100 mg
       831,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 1 170,00 €
    1 170,00 €
    1 * 200 mg
       1 170,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.