Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Absorbent Sheets
Absorbent Sheets
Supplier:   TRANSPOSAFE
Absorbent Sheets
Supplier:  TRANSPOSAFE
Certificates
Absorbent Sheets
4 Options Available Below
Product Details & Documents
The Transposafe absorption material is available in various models, sizes and absorption volumes. The absorption sheets offer various application options.
  • High absorption, suitable for Post NL-approved shipments
    Suitable for safely transporting blood and other diagnostic materials
    Can be used in the food industry
They can be used for the transport of fluids such as blood and other diagnostic materials that fall under UN3373 regulations. They are also suitable for the food industry, for packaging and transporting meat or fish, among other things.
Product Family Options
Product Information
  • Absorbency
    Colour
    Availability
    Price
  • TRSPDF110140
    Absorbent sheet, 110×140 mm
    Absorbent sheet, 110×140 mm
    200 ml (Absorption capacity)
    White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 231,00 €
    231,00 €
    1 * 1.000 ST
       231,00 €
  • TRSPDF150240B
    Absorbent sheet, 3 tubes, 185×125 mm + 40 mm flap
    Absorbent sheet, 3 tubes, 185×125 mm + 40 mm flap
    225 ml (Absorption capacity)
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
  • TRSPDF225250
    Absorbent sheet, 225×250 mm
    Absorbent sheet, 225×250 mm
    1 L (Absorption capacity)
    White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 543,00 €
    543,00 €
    1 * 1.000 ST
       543,00 €
  • 216-1613
    Absorbent sheet, 210×300 mm
    Absorbent sheet, 210×300 mm
    1,5 L (Absorption capacity)
    White
    Log in for availability
    Log in for availability
    1 * 375 ST
    1 * 375 ST - 195,00 €
    195,00 €
    1 * 375 ST
       195,00 €
    Specifications:
    New Product