Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

S14-95 ≥95% JAK/STAT pathway inhibitor

Catalog # ABCAAB144515-500
Marketsource
Supplier:   Abcam
S14-95 ≥95% JAK/STAT pathway inhibitor
Catalog # ABCAAB144515-500
Supplier:  Abcam
Marketsource
CAS Number:  419532-92-6
S14-95 ≥95% JAK/STAT pathway inhibitor
CAS Number:  419532-92-6
Price
441,00 €
Log in for availability
1 * 1 EA
Product Details & Documents

JAK/STAT pathway inhibitor. Inhibits IFN-γ mediated expression of the reporter gene (IC₅₀ values are 5,4 to 10,8 μM). Inhibits COX-2 and NOS II expression (IC₅₀ =10,8 μM). Inhibits iNOS-dependent NO production.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:419532-92-6
  • Molecular Weight:450,6 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% [(1R,2S,5S,7R,10R)-2,6,6,10-tetramethyl-16-oxo-14-phenyl-11,15-dioxatetracyclo[8,8,0,02,7,012,17]octadeca-12(17),13-dien-5-yl] acetate
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.