Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Melatonin ≥99% Endogenous hormone agonist at MT1 and MT2 receptors

Catalog # ABCAAB141052-50
Marketsource
Supplier:   Abcam
Melatonin ≥99% Endogenous hormone agonist at MT1 and MT2 receptors
Catalog # ABCAAB141052-50
Supplier:  Abcam
Marketsource
CAS Number:  73-31-4
Melatonin ≥99% Endogenous hormone agonist at MT1 and MT2 receptors
CAS Number:  73-31-4
Price
61,50 €
Log in for availability
1 * 50 mg
Product Details & Documents

Agonist at melatonin MT1 and MT2 receptors. Endogenous hormone important in the control of the circadian rhythm. Wide range of pathophysiological functions including antioxidative and immunomodulatory effects.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Molecular Weight:232,28 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Melatonin