Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Ca2+/Calmodulin Solution 10x, Abcam

Ca2+/Calmodulin Solution 10x
Catalog # ABCAAB189137-500
Marketsource
Supplier:   Abcam
Ca2+/Calmodulin Solution 10x, Abcam
Catalog # ABCAAB189137-500
Supplier:  Abcam
Marketsource
CAS Number:  
Ca2+/Calmodulin Solution 10x, Abcam

Ca2+/Calmodulin Solution 10x

CAS Number:  
Price
66,70 €
Log in for availability
1 * 500 µl
Product Details & Documents

10x stock solution used for assaying protein kinase activity of CAMK family members.

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Ca2+/Calmodulin Solution 10x
  • Cat. No.:
    ABCAAB189137-500
  • Storage temperature:
    Store at -20°C.

Specifications