Specifications
- Description:LEDlight meter for apollo II liquid viewer
- Seq:1
Specifications
About this item
Please note that this listing only refers to the LED Light Meter, which accompanies the Apollo II liquid viewer. Designed for use in pharmaceutical and laboratory environments, the Apollo II liquid viewer for transparent glass containers provides a simple method for the detection of visible particles in ampoules, vials, and bottles.
Height and angle adjustable
- Intuitive assembly; no tools required
- Light level between 2,000 and 3,750 Lux
- Unit can fold for convenient storage, with the inspection surfaces folding together to reduce chance of damage
The unit is packaged in sustainably sourced foam, made from polyethylene resin which is derived from renewable sugarcane-based feedstock.
The Apollo II liquid viewer is suitable for transparent glass containers. Adelphi Manufacturing also offer the Apollo II high intensity liquid viewer for coloured glass containers, and the Apollo I liquid viewer, with additional magnifier.
This unit conforms to the European Pharmacopoeia Specification 2.9.20 and to the US Pharmacopoeia – USP chapter 41 (1790), section 6.1. Adelphi Manufacturing’s ISO 9001:2015 certification guarantees the high quality finish of this product.