Order Entry
Netherlands
ContactUsLinkComponent
Enterococcus faecalis WDCM 00087 Vitroids™, Certified Reference Material, Suitable for Microbiology
Enterococcus faecalis WDCM 00087 Vitroids™, Certified Reference Material, Suitable for Microbiology
Catalog # MILXVT000876-10EA
Supplier:  Merck
New Product
CAS Number:  
Enterococcus faecalis WDCM 00087 Vitroids™, Certified Reference Material, Suitable for Microbiology
Catalog # MILXVT000876-10EA
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Standards:
    Certified reference material
  • Description:
    Enterococcus faecalis WDCM 00087 Vitroids™, 1000 - 10000 CFU mean value range
  • Storage conditions:
    −20 °C
  • Product type:
    Solid

Specifications

About this item

Enterococcus faecalis WDCM 00087 Vitroids are certified microorganisms standards (Certified Microbiological Reference Materials), which can be used as quantitative and qualitative control organisms. They contain colony-forming units (CFUs) in the range of 1000 to 10000 CFU per disc.

  • Ready-to-use at convenient CFU ranges

  • No dilution required
  • Quick rehydration in 10 minutes
  • Two to three years of shelf life in a standard −20 °C freezer
  • Complete with a comprehensive certificate of analysis
  • No maintenance of stock cultures
  • No recovery time or pre-enrichment step is necessary
  • Robust, efficient, and reliable
  • Easy-to-use and cost-effective

The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms.

 

These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from CECT®. These ready-to-use microbiological controls minimize the need for maintaining control strains in the test laboratory and guarantee the use of an authenticated control culture for every quality control test.

Vitroids are derived from a traceable culture obtained freeze-dried from [TM='CECT'].

Application:

Enterococcus faecalis WDCM 00087 Vitroids are used as reference strains in routine quality control for food, beverage, water, and environmental testing. Additionally, they are excellent for use in method development and validations, as starter cultures, and for use in research studies and educational purposes.

Packaging:

Package of 10 discs.