Order Entry
Netherlands
ContactUsLinkComponent
Gonyautoxin-2 and Gonyautoxin-3 Solution
Gonyautoxin-2 and Gonyautoxin-3 Solution
Catalog # 55739-0.5ML
Supplier:  Merck
New Product
CAS Number:  
Gonyautoxin-2 and Gonyautoxin-3 Solution
Catalog # 55739-0.5ML
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Concentration:
    20 μg/g in acetic acid
  • Description:
    Gonyautoxin-2 and Gonyautoxin-3 solution, 0,5 ml

Specifications

About this item

For use in the analysis of food and cosmetics and other analytical applications, we offer a comprehensive portfolio comprised of thousands of standards and Certified Reference Materials (CRMs).

Our high-purity standards help to keep the food and cosmetic products safe and allow the manufacturers and laboratories to meet all legal regulations for label claims.