Order Entry
Netherlands
ContactUsLinkComponent
 
Water ≥99.9%, LiChrosolv®, PFAS grade, Supelco®
  1047351000.
 :  Merck
New Product
   Safer Human and Environmental Health
 :  
undefined
Water ≥99.9%, LiChrosolv®, PFAS grade, Supelco®
  1047351000.
 :  Merck
New Product
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Borosilicate glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

 

 

  • EPA 533/537.1 Tested:
    Conforms
  • UHPLC Gradient (at 230 nm):
    ≤2.0 mAU
  • Evaporation Residue:
    ≤0.7 mg/l
  • Titratable Acid (meq/g):
    ≤0.001
  • Titratable Base (meq/g):
    ≤0.0002

 

 

 : Hydrogen oxide
 :
 :18,02 g/mol
 :Room Temperature
 :7732-18-5
 :H₂O
 :MFCD00011332
 :100,0 °C at 1.013 hPa
 :0 °C
 :1,000 g/cm³
 :M-Clarity™ quality level = MQ100