Order Entry
Netherlands
ContactUsLinkComponent
Rat Tissue Plasminogen Activator ELISA Kit (TPA)
Rat Tissue Plasminogen Activator ELISA Kit (TPA)
Catalog # ABCAAB198510-96
Supplier:  Abcam
CAS Number:  
undefined
Rat Tissue Plasminogen Activator ELISA Kit (TPA)
Catalog # ABCAAB198510-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Rat Tissue Plasminogen Activator ELISA kit (TPA)
  • Tests per kit:
    96 tests
Specifications
About this item
Rat Tissue Plasminogen Activator ELISA Kit (TPA) is a Quantitative ELISA kit for the measurement of Rat Tissue Plasminogen Activator (TPA) in Rat in Plasma, Other biological fluids samples.