Mouse mTOR ELISA Kit
ABCAAB206311-96
: Abcam
:
- Description:Mouse mTOR ELISA kit
- Tests per kit:96 tests
Mouse mTOR ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse mTOR with a sensitivity of 16,7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.
: RUO