Human TNF alpha ELISA Set (without plates)
Supplier: Abcam
Certificates
About this item
Human TNF alpha ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human TNF alpha ELISA Set (without plates) in Human in plasma, cell culture supernatant, serum samples.
Specifications
- Storage temperature:+4 ℃