Human Complement Factor P ELISA Kit
Catalog # ABCAAB222864-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Complement Factor P ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Complement Factor P ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human Complement Factor P in Human in Cerebral Spinal Fluid, Saliva, Urine, plasma, serum samples.