Order Entry
Netherlands
ContactUsLinkComponent
Inositol Triphosphate (IP3) ELISA Kit
Inositol Triphosphate (IP3) ELISA Kit
Catalog # ABCAAB287832-96
Supplier:  Abcam
CAS Number:  
undefined
Inositol Triphosphate (IP3) ELISA Kit
Catalog # ABCAAB287832-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Inositol Triphosphate (IP3) ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Inositol Triphosphate (IP3) ELISA Kit is a Competitive ELISA kit for the measurement of Inositol Triphosphate (IP3) in Plasma, Serum, Other biological fluids samples.