Human Granzyme B ELISPOT Set
: Abcam
Human Granzyme B ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Granzyme B production in a single cell suspension.
- Species reactivity:Human
- Sample type:Suspension cells
- Form:Uncoated
- Storage temperature:+4 °C
- Processing time:Multiple steps
- Antigen symbol:Granzyme B
- Regulatory status:RUO
Frequently Bought Together