Rat Cystatin C ELISA Kit
Catalog # ABCAAB201281-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Cystatin C ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rat Cystatin C ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat Cystatin C with a sensitivity of 11,72 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 5 citations.