Mouse Endocan ELISA Kit
Catalog # ABCAAB236716-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Endocan ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse Endocan ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Endocan with a sensitivity of 9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.