Rabbit Plasminogen ELISA Kit
Catalog # ABCAAB190544-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rabbit Plasminogen ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Rabbit Plasminogen ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit Plasminogen in Rabbit in Plasma, Serum samples.