Mouse Fibronectin ELISA Kit
Catalog # ABCAAB108849-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Fibronectin ELISA kit
Specifications
About this item
Mouse Fibronectin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Fibronectin in Mouse in urine, plasma, serum samples.