Order Entry
Netherlands
ContactUsLinkComponent
Pyruvate dehydrogenase (PDH) Profiling ELISA Kit
Pyruvate dehydrogenase (PDH) Profiling ELISA Kit
Catalog # ABCAAB110174-96
Supplier:  Abcam
CAS Number:  
undefined
Pyruvate dehydrogenase (PDH) Profiling ELISA Kit
Catalog # ABCAAB110174-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Pyruvate dehydrogenase (PDH) Profiling ELISA kit
  • Storage temperature:
    Dual Temp (Dry Ice)
  • Tests per kit:
    96 tests

Specifications

About this item

Pyruvate dehydrogenase (PDH) Profiling ELISA kit is a Sandwich (qualitative) ELISA kit for the measurement of Pyruvate dehydrogenase (PDH) Profiling in cell culture extracts, tissue samples from a wide range of species.