Order Entry
Netherlands
ContactUsLinkComponent
Human beta 2 Microglobulin ELISA Kit (B2M)
Human beta 2 Microglobulin ELISA Kit (B2M)
Catalog # ABCAAB99977-96
Supplier:  Abcam
CAS Number:  
undefined
Human beta 2 Microglobulin ELISA Kit (B2M)
Catalog # ABCAAB99977-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Human beta 2 Microglobulin ELISA Kit (B2M)
  • Tests per kit:
    96 tests
Specifications
About this item
Human beta 2 Microglobulin ELISA Kit (B2M) is a Sandwich (quantitative) ELISA kit for the measurement of Human beta 2 Microglobulin (B2M) in Human in Plasma, Cell culture supernatant, Serum samples.