Human Fractalkine ELISA Kit (CX3CL1)
Catalog # ABCAAB100522-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Fractalkine ELISA kit (CX3CL1)
- Tests per kit:96 tests
Specifications
About this item
Human Fractalkine ELISA Kit (CX3CL1) is a Sandwich (quantitative) ELISA kit for the measurement of Human Fractalkine (CX3CL1) in Human in Plasma, Cell culture supernatant samples.