Anti-Kallikrein 6 Rabbit Monoclonal Antibody [clone: EPR23352-86]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR23352-86] to Kallikrein 6 - BSA and Azide free (Capture).
Type: Primary
Antigen: Kallikrein 6
Clonality: Monoclonal
Clone: EPR23352-86
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together