Anti-CXCL13 Rabbit Monoclonal Antibody [clone: EPR23400-92]
: Abcam
New Product
Rabbit monoclonal [EPR23400-92] to CXCL13.
Type: Primary
Antigen: CXCL13
Clonality: Monoclonal
Clone: EPR23400-92
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together