Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD3 Rat Monoclonal Antibody [clone: CD3-12] FITC (Fluorescein Isothiocyanate)
Anti-CD3 Rat Monoclonal Antibody [clone: CD3-12] FITC (Fluorescein Isothiocyanate)
Catalog # ABCAAB34722-50
Supplier:  Abcam
undefined
Anti-CD3 Rat Monoclonal Antibody [clone: CD3-12] FITC (Fluorescein Isothiocyanate)
Catalog # ABCAAB34722-50
Supplier:  Abcam

Specifications

  • Size:
    50 µg
  • Pack type:
    Vial

Specifications

About this item

FITC Rat monoclonal [CD3-12] to CD3.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: CD3-12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human; Mouse; Chicken; Dog; Horse; Monkey; Pig; Rhesus Monkey