Anti-Surfactant protein D/SP-D Rabbit Monoclonal Antibody [clone: EPR21928-91]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR21928-91] to Surfactant protein D/SP-D - BSA and Azide free (Detector).
Type: Primary
Antigen: Surfactant protein D/SP-D
Clonality: Monoclonal
Clone: EPR21928-91
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together