Anti-Niemann Pick C2 Rabbit Monoclonal Antibody [clone: EPR19993-145-1]
: Abcam
New Product
Rabbit monoclonal [EPR19993-145-1] to Niemann Pick C2.
Type: Primary
Antigen: Niemann Pick C2
Clonality: Monoclonal
Clone: EPR19993-145-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together